Learn More
Genscript Corporation NKG2D/CD314 hFc Chimera Flag
Catalog No. 5000056928
Encompass
NKG2D is a type II transmembrane glycoprotein having an extracellular lectin-like domain This domain lacks the recognizable calcium-binding sites found in true C-type lectins and binds protein rather than carbohydrate ligands Human NKG2D is expressed on CD8 alpha beta T cells gamma T cells NK cells and NKT cells
Catalog No. 50-000-56928 Supplier Genscript Corporation Supplier No. Z04278100
NKG2D is a type II transmembrane glycoprotein having an extracellular lectin-like domain This domain lacks the recognizable calcium-binding sites found in true C-type lectins and binds protein rather than carbohydrate ligands Human NKG2D is expressed on CD8 alpha beta T cells gamma T cells NK cells and NKT cells