Learn More
Genscript Corporation NKG2D/CD314 hFc Chimera Mouse
Catalog No. 5000059464
Encompass
NKG2D is a type II transmembrane glycoprotein having an extracellular lectin-like domain This domain lacks the recognizable calcium-binding sites found in true C-type lectins and binds protein rather than carbohydrate ligands Human NKG2D is expressed on CD8 alpha beta T cells gamma T cells NK cells and NKT cells
Catalog No. 50-000-59464 Supplier Genscript Corporation Supplier No. Z04345100
NKG2D is a type II transmembrane glycoprotein having an extracellular lectin-like domain This domain lacks the recognizable calcium-binding sites found in true C-type lectins and binds protein rather than carbohydrate ligands Human NKG2D is expressed on CD8 alpha beta T cells gamma T cells NK cells and NKT cells