Learn More
ACROBiosystems ACROBiosystems Human NKG2D / CD314 Protein, Fc Tag (HPLC-verified)
Catalog No. 501956713
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2D, Fc Tag (HPLC-verified) (NKD-H5265) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 43.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human MICA, His Tag (Cat. No. MIA-H5221) at 5 μg/mL (100 μL/well) can bind Human NKG2D, Fc Tag (HPLC-verified) (Cat. No. NKD-H5265) with a linear range of 4-63 ng/mL (QC tested).
Catalog No. 50-195-6713 Supplier ACROBiosystems Supplier No. NKDH5265100UG
Recombinant Protein;100UG;Human NKG2D, Fc Tag (HPLC-verified) (NKD-H5265) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 43.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human MICA, His Tag (Cat. No. MIA-H5221) at 5 μg/mL (100 μL/well) can bind Human NKG2D, Fc Tag (HPLC-verified) (Cat. No. NKD-H5265) with a linear range of 4-63 ng/mL (QC tested).