Learn More
ACROBiosystems ACROBiosystems Human NKG2D / CD314 Protein, His Tag
Catalog No. 501956607
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.5 kDa.
Catalog No. 50-195-6607 Supplier ACROBiosystems Supplier No. NKDH5245100UG
Recombinant Protein;100UG;Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.5 kDa.