Learn More
ACROBiosystems ACROBiosystems Human NKG2D / CD314 Protein, His Tag
Supplier:  ACROBiosystems NKDH52451MG
Encompass_Preferred
Recombinant Protein;1MG;Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.5 kDa.
Catalog No. 50-195-6606