Learn More
Epigentek Inc NKG7 Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A60026100
Encompass_Preferred
Biotin Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Protein NKG7 protein (30-60AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Protein NKG7, NKG7, GIG1, G-CSF-induced gene 1 protein, GIG-1 protein, Granule membrane protein of 17 kDa, GMP-17, Natural killer cell protein 7, p15-TIA-1.
Catalog No. 50-123-4126