Learn More
Abcam NKH 477 (Colforsin daropate), an adenylyl cyclase activator, 10MG
Catalog No. 502707806
Encompass_Preferred
MW 546.1 Da. Water-soluble adenylyl cyclase activator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7806 Supplier Abcam Supplier No. AB12019010MG
Add to Cart
Add to Cart
MW 546.1 Da. Water-soluble adenylyl cyclase activator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement