Learn More
Cayman Chemical NKH477hydrochlorIde 10mg
SDP
Catalog No. 502951338
Encompass_Preferred
A water-soluble analog of forskolin which has both inotropic and vasodilator effects when administered intravenously; stimulates cardiac (type V) adenylyl cyclase more potently than other isoforms; relaxes guinea pig tracheal smooth muscle precontracted with histamine with an EC50 value of 32.6 nM
Catalog No. 50-295-1338 Supplier Cayman Chemical Supplier No. 1121410 MG
A water-soluble analog of forskolin which has both inotropic and vasodilator effects when administered intravenously; stimulates cardiac (type V) adenylyl cyclase more potently than other isoforms; relaxes guinea pig tracheal smooth muscle precontracted with histamine with an EC50 value of 32.6 nM