Learn More
Aviva Systems Biology Corp NKIRAS1 Antibody - N-terminal region (ARP55370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKIRAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NKIRAS1. Alternate names: KBRAS1, kappaB-Ras1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61481