Learn More
Medchemexpress LLC HY-100384 10mg Medchemexpress, NKL 22 CAS:537034-15-4 Purity:>98%
SDP
Catalog No. 502080585
Encompass_Preferred
Medchemexpress, HY-100384 10mg NKL 22 CAS:537034-15-4 NKL 22 (compound 4b) is a potent and selective inhibitor of histone deacetylases (HDAC), with an IC50 of 199 and 69 nM for HDAC1 and HDAC3, respectively. NKL 22 exhibits selectivity over HDAC2/4/5/7/8 (IC50≥1.59 μM). NKL 22 ameliorates the disease phenotype and transcriptional abnormalities in Huntington's disease transgenic mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0585 Supplier Medchemexpress LLC Supplier No. HY10038410MG
Medchemexpress, HY-100384 10mg NKL 22 CAS:537034-15-4 NKL 22 (compound 4b) is a potent and selective inhibitor of histone deacetylases (HDAC), with an IC50 of 199 and 69 nM for HDAC1 and HDAC3, respectively. NKL 22 exhibits selectivity over HDAC2/4/5/7/8 (IC50≥1.59 μM). NKL 22 ameliorates the disease phenotype and transcriptional abnormalities in Huntington's disease transgenic mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.