Learn More
Medchemexpress LLC HY-16350 5mg Medchemexpress, NKP-1339 CAS:197723-00-5 Purity:>98%
SDP
Catalog No. 501932769
Encompass_Preferred
Medchemexpress, HY-16350 5mg NKP-1339 CAS:197723-00-5 NKP-1339 (IT-139) is the first-in-class ruthenium-based anticancer drug in clinical development against solid cancer with limited side effects. NKP-1339 induces G2/M cell cycle arrest, blockage of DNA synthesis , and induction of apoptosis via the mitochondrial pathway. NKP-1339 has a high tumor targeting potential, strongly binds to serum proteins such as albumin and transferrin and activates in the reductive tumor milieu [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2769 Supplier Medchemexpress LLC Supplier No. HY163505MG
Medchemexpress, HY-16350 5mg NKP-1339 CAS:197723-00-5 NKP-1339 (IT-139) is the first-in-class ruthenium-based anticancer drug in clinical development against solid cancer with limited side effects. NKP-1339 induces G2/M cell cycle arrest, blockage of DNA synthesis , and induction of apoptosis via the mitochondrial pathway. NKP-1339 has a high tumor targeting potential, strongly binds to serum proteins such as albumin and transferrin and activates in the reductive tumor milieu [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.