Learn More
ACROBiosystems Recombinant Protein;Human NKp30 / NCR3 / CD337 Protein, Fc Tag;HEK293;1MG;NC3,NCR3,CD337,NKp30,NK-p30,1C7,LY117,MALS,NCR-3
Catalog No. 501658463
Encompass_Preferred
Recombinant Protein;Human NKp30 / NCR3 / CD337 Protein, Fc Tag;1MG;Human NKp30, Fc Tag (NC3-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.5 kDa. As a result of glycosylation, the protein migrates as 45-55 kDa under reducing (R) condition, and 90-110 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8463 Supplier ACROBiosystems Supplier No. NC3H52591MG
Recombinant Protein;Human NKp30 / NCR3 / CD337 Protein, Fc Tag;1MG;Human NKp30, Fc Tag (NC3-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.5 kDa. As a result of glycosylation, the protein migrates as 45-55 kDa under reducing (R) condition, and 90-110 kDa under non-reducing (NR) condition (SDS-PAGE).