Learn More
ACROBiosystems Human NKp30 Protein, His Tag, 100 μg
Catalog No. 501658458
Encompass_Preferred
This human NKp30 (NCR3, CD337) protein is a member of the natural cytotoxicity receptor family, containing one Ig-like domain. It is expressed in HEK293 cells and carries a C-terminal polyhistidine tag. The protein plays a critical role in NK cell activation and dendritic cell maturation through its interaction with ligands such as BAG6.
  • Greater than 95% purity determined by SDS-PAGE
  • Calculated molecular weight of 13.7 kDa, migrating as 21-32 kDa under reducing conditions due to glycosylation
  • Endotoxin level less than 1.0 EU per μg
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-165-8458 Supplier ACROBiosystems Supplier No. NC3H5228100UG
This human NKp30 (NCR3, CD337) protein is a member of the natural cytotoxicity receptor family, containing one Ig-like domain. It is expressed in HEK293 cells and carries a C-terminal polyhistidine tag. The protein plays a critical role in NK cell activation and dendritic cell maturation through its interaction with ligands such as BAG6.
  • Greater than 95% purity determined by SDS-PAGE
  • Calculated molecular weight of 13.7 kDa, migrating as 21-32 kDa under reducing conditions due to glycosylation
  • Endotoxin level less than 1.0 EU per μg
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state