Learn More
Enzo Life Sciences NKp44/NCR2 monoclonal antibody (AT1G6) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7340100
Encompass_Preferred
Clone: AT1G6. Isotype: IgG2aκ. Immunogen: Recombinant human NKp44 (aa 19-130) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3275