Learn More
ACROBiosystems ACROBiosystems Biotinylated Human NKp46 / NCR1 / CD335 Protein, Fc,Avitag™
Supplier:  ACROBiosystems NC1H82F9200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.7 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6634