Learn More
ACROBiosystems ACROBiosystems Human NKp46 / NCR1 / CD335 Protein, His Tag
Catalog No. 501956680
Encompass_Preferred
Recombinant Protein;100UG;Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.3 kDa. The protein migrates as 36-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6680 Supplier ACROBiosystems Supplier No. NC1H52H4100UG
Recombinant Protein;100UG;Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.3 kDa. The protein migrates as 36-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.