Learn More
ACROBiosystems ACROBiosystems Human NKp46 / NCR1 / CD335 Protein, Llama IgG2b Fc Tag, low endotoxin
Supplier:  ACROBiosystems NC1H5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human NKp46, Llama IgG2b Fc Tag, low endotoxin (NC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6647