Learn More
Enzo Life Sciences NKp46/NCR1 monoclonal antibody (N1D9) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7360100
Encompass_Preferred
Clone: n1D9. Isotype: IgG1κ. Immunogen: Recombinant human Nkp46/NCR1 (aa 22-255) purified from E. coli. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ELISA, Flow Cytometry. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3277