Learn More
ACROBiosystems Recombinant Protein;Human NKp46 / NCR1 / CD335 Protein, Fc Tag;HEK293;1MG;NC1,NCR1,LY94,CD335,NK-p46,NKp46,hNKp46,NCR-1,LY-94
Catalog No. 501658515
Encompass_Preferred
Recombinant Protein;Human NKp46 / NCR1 / CD335 Protein, Fc Tag;1MG;Human NKp46, Fc Tag (NC1-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53 kDa. The protein migrates as 53-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8515 Supplier ACROBiosystems Supplier No. NC1H52571MG
Recombinant Protein;Human NKp46 / NCR1 / CD335 Protein, Fc Tag;1MG;Human NKp46, Fc Tag (NC1-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53 kDa. The protein migrates as 53-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.