Learn More
Aviva Systems Biology Corp NKX1-1 Antibody - middle region (ARP37637_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37637_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX1-1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse NKX1-1. Alternate names: Sax2, Nkx-1.1ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53086