Learn More
Aviva Systems Biology Corp Nkx1-1 Antibody - middle region (ARP37638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nkx1-1. It was validated on Western Blot. Nkx1-1,NK1 transcription factor related, locus 1 (Drosophila),Sax2; Nkx-1.1; Nkx1-1,NK1 transcription factor related, locus 1 (Drosophila)The immunogen is a synthetic peptide directed towards the middle region of human Nkx1-1. Alternate names: Nkx-1.1, Sax2ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-284-674