Learn More
Aviva Systems Biology Corp NKX1-1 Antibody - middle region (ARP47803_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47803_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX1-1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NKX1-1. Alternate names: HSPX153, SAX2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59669