Learn More
Aviva Systems Biology Corp Nkx1-2 Antibody - middle region (ARP36974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nkx1-2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Nkx1-2. Alternate names: Nkx-1.1, Nkx-1.2, Sax1ApplicationWB. Species reactivity: Mouse, Pig, Rat
Catalog No. 50-284-556