Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4821MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM564. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: Pancreas or Ewing sarcoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human NKX2.2 recombinant protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40-50kDa.Chromosome: 20p11.22. Gene Symbol: NKX2-2. Gene ID: 4821. Ensemble: ENSG00000125820. Uniprot: XP_006723629.
Catalog No. 50-188-5019