Learn More
Aviva Systems Biology Corp NKX2-2 Antibody - N-terminal region (ARP32338_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32338_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NKX2-2. Alternate names: NKX2.2, NKX2BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54532