Learn More
Aviva Systems Biology Corp NKX2-2 Antibody - N-terminal region (ARP37110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Nkx2.2, tinman, Nkx-2.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53065