Learn More
Aviva Systems Biology Corp NKX2-3 Antibody - middle region (ARP36943_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36943_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse NKX2-3. Alternate names: Nkx2.3, tinman, Nkx-2.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53063