Learn More
Aviva Systems Biology Corp Nkx2-3 Antibody - C-terminal region (ARP36944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nkx2-3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the c terminal region of mouse Nkx2-3. Alternate names: Nkx-2.3, Nkx2.3, tinmanApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-62885