Learn More
Aviva Systems Biology Corp NKX2-4 Antibody - C-terminal region (ARP37326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse NKX2-4. Alternate names: tinman, Nkx-2.4, 1700001P03RikApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-53069