Learn More
Aviva Systems Biology Corp Nkx2-5 Antibody - N-terminal region (ARP31575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human Nkx2-5. Alternate names: CSX, CSX1, NKX2.5, NKX2E, NKX4-1, VSD3, CHNG5, HLHS2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54271