Learn More
Aviva Systems Biology Corp NKX2-8 Antibody - C-terminal region (ARP31856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX2-8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NKX2-8. Alternate names: NKX2.8, NKX2H, Nkx2-9ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54342