Learn More
Biocare Medical, LLC NKX3.1, Human Protein, 1.0 ml, Stains Nuclei in Both Normal and Prostate Cancer
SDP
Supplier:  Biocare Medical, LLC CP422A
Encompass_Preferred
NKX3.1 is positive in the vast majority of primary prostatic adenocarcinomas. It is 98.6% sensitive for metastatic prostatic adenocarcinomas. Specificity was 99.7% in various cancers and it stains nuclei in both normal and prostate cancer.
Catalog No. 50-827-88