Learn More
Aviva Systems Biology Corp NKX3-1 Antibody - middle region (ARP38751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX3-1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NKX3-1. Alternate names: BAPX2, NKX3, NKX3.1, NKX3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56782