Learn More
Aviva Systems Biology Corp Nkx3-2 Antibody - middle region (ARP36827_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nkx3-2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Nkx3-2. Alternate names: Bapx1, NKX3.2, Nkx-3.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Mouse, Rabbit, Rat
Catalog No. 50-284-518