Learn More
Aviva Systems Biology Corp NKX3-2 Antibody - middle region (ARP58064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX3-2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NKX3-2. Alternate names: BAPX1, MGC138171, NKX3.2, NKX3B, SMMDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-62227