Learn More
Aviva Systems Biology Corp NKX6-3 Antibody - N-terminal region (ARP39906_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39906_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NKX6-3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NKX6-3. Alternate names: NKX6.3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53159