Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nky80 | 299442-43-6 | MFCD02026323 | 99.8% | 229.23 g/mol | C12H11N3O2 | 10 MG
SDP

Catalog No. 5000246002
Encompass_Preferred

NKY80 (CAS 299442-43-6) is a small-molecule quinazolinone derivative that selectively and non-competitively inhibits adenylyl cyclase type V. It is used in biochemical and cellular research to probe cAMP signaling pathways and is supplied as a high-purity solid suitable for in vitro assays and for preparation of in vivo formulations with appropriate solvents and carriers.

  • High purity: 99.8%
  • Molecular weight: 229.23 g/mol
  • Chemical formula: C12H11N3O2
  • Solubility: soluble in DMSO (≈250 mg/mL)
  • Applications: in vitro enzyme and cellular assays
  • Storage: powder at -20°C for long-term stability

Catalog No. 50-002-46002 Supplier Medchemexpress LLC Supplier No. HY10319510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NKY80 (CAS 299442-43-6) is a small-molecule quinazolinone derivative that selectively and non-competitively inhibits adenylyl cyclase type V. It is used in biochemical and cellular research to probe cAMP signaling pathways and is supplied as a high-purity solid suitable for in vitro assays and for preparation of in vivo formulations with appropriate solvents and carriers.

  • High purity: 99.8%
  • Molecular weight: 229.23 g/mol
  • Chemical formula: C12H11N3O2
  • Solubility: soluble in DMSO (≈250 mg/mL)
  • Applications: in vitro enzyme and cellular assays
  • Storage: powder at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.