Learn More
ATCC NL20; Bronchial Epithelium; Human (Homo sapiens)
Catalog No. 502382464
Encompass
NL20 is an epithelial cell line that was isolated from the normal bronchus of a White, 20-year-old, female patient. It is immortalized and nontumorigenic it can be used to study tumor progression along with NL20-TA (ATCC CRL-2504). This cell line was deposited by JH Schiller.
Catalog No. 50-238-2464 Supplier ATCC Supplier No. CRL2503
NL20 is an epithelial cell line that was isolated from the normal bronchus of a White, 20-year-old, female patient. It is immortalized and nontumorigenic it can be used to study tumor progression along with NL20-TA (ATCC CRL-2504). This cell line was deposited by JH Schiller.