Learn More
New England Biolabs, Inc. NlaIII – 500 units
SDP
Catalog No. 50811971
Encompass
A restriction endonuclease that recognizes the sequence _CATG^.
Catalog No. 50-811-971 Supplier New England Biolabs, Inc. Supplier No. R0125S
A restriction endonuclease that recognizes the sequence _CATG^.