Learn More
New England Biolabs, Inc. NlaIV – 200 units
SDP
Catalog No. 50811973
Encompass
A restriction endonuclease that recognizes the sequence GGN_^NCC.
Catalog No. 50-811-973 Supplier New England Biolabs, Inc. Supplier No. R0126S
A restriction endonuclease that recognizes the sequence GGN_^NCC.