Learn More
Apexbio Technology LLC NLE4-D-PHE7-945.-MSH-5MG
SDP
Catalog No. 5000569162
Encompass_Preferred
[Nle4 D-Phe7]- 945 -MSH (CAS No 75921-69-6) is a synthetic peptide analog of the naturally occurring alpha-melanocyte-stimulating hormone ( -MSH) studied in the fields of endocrinology and dermatology It binds primarily to melanocortin receptors playing a role in reducing inflammation and influencing pigmentation The compound exhibits potent activity in the nanomolar to low micromolar range during in vitro assays It is commonly used in cellular models to study melanocyte function and in animal studies focusing on skin pigmentation disorders and anti-inflammatory responses Moreover it serves as a valuable tool in drug screening for potential therapeutic agents targeting receptor pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69162 Supplier Apexbio Technology LLC Supplier No. B53635
[Nle4 D-Phe7]- 945 -MSH (CAS No 75921-69-6) is a synthetic peptide analog of the naturally occurring alpha-melanocyte-stimulating hormone ( -MSH) studied in the fields of endocrinology and dermatology It binds primarily to melanocortin receptors playing a role in reducing inflammation and influencing pigmentation The compound exhibits potent activity in the nanomolar to low micromolar range during in vitro assays It is commonly used in cellular models to study melanocyte function and in animal studies focusing on skin pigmentation disorders and anti-inflammatory responses Moreover it serves as a valuable tool in drug screening for potential therapeutic agents targeting receptor pathways Experimental concentrations typically depend on the specific research design