Learn More
Cayman Chemical Nle4 D-Phe7-a-MSHacetate 10mg
SDP
Catalog No. 502942951
Encompass_Preferred
A peptide agonist of MC1R; induces cAMP accumulation in mouse L-cells expressing human MC1R (EC50 = 0.5 nM); induces tyrosinase activity in mouse melanoma cells; induces long-term skin darkening in frogs at 1.8 mg/kg
Catalog No. 50-294-2951 Supplier Cayman Chemical Supplier No. 4154810 MG
A peptide agonist of MC1R; induces cAMP accumulation in mouse L-cells expressing human MC1R (EC50 = 0.5 nM); induces tyrosinase activity in mouse melanoma cells; induces long-term skin darkening in frogs at 1.8 mg/kg