Learn More
Cayman Chemical [Nle4,D-Phe7]-α-MSH (acetate), Purity: ≥98%, CAS: 1566590-77-9, 5 mg
SDP
Catalog No. 502942921
Encompass_Preferred
A peptide agonist of MC1R; induces cAMP accumulation in mouse L-cells expressing human MC1R (EC50 = 0.5 nM); induces tyrosinase activity in mouse melanoma cells; induces long-term skin darkening in frogs at 1.8 mg/kg
Catalog No. 50-294-2921 Supplier Cayman Chemical Supplier No. 415485 MG
A peptide agonist of MC1R; induces cAMP accumulation in mouse L-cells expressing human MC1R (EC50 = 0.5 nM); induces tyrosinase activity in mouse melanoma cells; induces long-term skin darkening in frogs at 1.8 mg/kg