Learn More
CPC Scientific H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PTHP003B
Encompass
ONE-LETTER SEQUENCE: SVSEIQL-Nle-HNLGKHLNS-Nle-ERVEWLRKKLQDVHNYMOLECULAR FORMULA: C183H295N55O52MOLECULAR WEIGHT:4097.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [78041-34-6], RESEARCH AREA: OsteoporosisREFERENCES: T. Noda et al., 41st Chem. Soc. of Japan Meeting, 4S12, Osaka, Japan (Apr. 1980)
Catalog No. 50-211-0338