Learn More
CPC Scientific H-Ser-Glu-Ile-Gln-Leu-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110339
Encompass
ONE-LETTER SEQUENCE: SEIQL-Nle-HNLGKHLNS-Nle-ERVEWLRKKLQDVHNY-NH2MOLECULAR FORMULA: C175H282N54O48MOLECULAR WEIGHT:3910.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [120527-89-1], SYNONYMS: [Nle8,18,Tyr34]-Parathyroid Hormone (Human, 3-34 Amide)RESEARCH AREA: Osteoporosis
Catalog No. 50-211-0339 Supplier CPC Scientific Supplier No. PTHP004A
ONE-LETTER SEQUENCE: SEIQL-Nle-HNLGKHLNS-Nle-ERVEWLRKKLQDVHNY-NH2MOLECULAR FORMULA: C175H282N54O48MOLECULAR WEIGHT:3910.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [120527-89-1], SYNONYMS: [Nle8,18,Tyr34]-Parathyroid Hormone (Human, 3-34 Amide)RESEARCH AREA: Osteoporosis