Learn More
Cayman Chemical nlfInavIrmesylate 10mg
SDP
Catalog No. 502969021
Encompass_Preferred
An orally bioavailable inhibitor of the HIV-1 protease (Ki =2 nM) that is used in combination with HIV reverse transcriptase inhibitors for the treatment of HIV infection
Catalog No. 50-296-9021 Supplier Cayman Chemical Supplier No. 1514410 MG
An orally bioavailable inhibitor of the HIV-1 protease (Ki =2 nM) that is used in combination with HIV reverse transcriptase inhibitors for the treatment of HIV infection