Learn More
Cayman Chemical Nelfinavir (mesylate), Purity: ≥98%, CAS: 159989-65-8, 50 mg
SDP
Catalog No. 502970786
Encompass_Preferred
An orally bioavailable inhibitor of the HIV-1 protease (Ki =2 nM) that is used in combination with HIV reverse transcriptase inhibitors for the treatment of HIV infection
Catalog No. 50-297-0786 Supplier Cayman Chemical Supplier No. 1514450 MG
An orally bioavailable inhibitor of the HIV-1 protease (Ki =2 nM) that is used in combination with HIV reverse transcriptase inhibitors for the treatment of HIV infection