Learn More
Aviva Systems Biology Corp NLGN4X Antibody - middle region (ARP49557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NLGN4X. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NLGN4X. Alternate names: ASPGX2, AUTSX2, HLNX, HNLX, KIAA1260, MGC22376, NLGN, NLGN4, HNL4XApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60112