Learn More
Cell Signaling Technology NLK Monoclonal Antibody, Rabbit, Human, Mouse, Rat, 100 μL
SDP
Catalog No. 502729905
Encompass
The NLK (D9X3C) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total NLK protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human NLK protein. NLK is a serine/threonine-protein kinase that regulates signaling pathways including Wnt/β-catenin, TGFβ, IL-6, and Notch, and plays a role in cell proliferation, differentiation, and nervous system development.
  • Validated for western blotting applications.
  • Demonstrates reactivity with human, mouse, and rat species.
  • Optimized for endogenous sensitivity.
  • Supplied in a formulation containing 50% glycerol.
  • Recommended dilution for western blot is 1:1,000.
Catalog No. 50-272-9905 Supplier Cell Signaling Technology Supplier No. 94350S
The NLK (D9X3C) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total NLK protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human NLK protein. NLK is a serine/threonine-protein kinase that regulates signaling pathways including Wnt/β-catenin, TGFβ, IL-6, and Notch, and plays a role in cell proliferation, differentiation, and nervous system development.
  • Validated for western blotting applications.
  • Demonstrates reactivity with human, mouse, and rat species.
  • Optimized for endogenous sensitivity.
  • Supplied in a formulation containing 50% glycerol.
  • Recommended dilution for western blot is 1:1,000.