Learn More
Bioworld NLN Medium w/ Vitamins, 25 L
Catalog No. 50254760
Encompass_Preferred
NLN MediumContains Macronutrients, Micronutrients, and Vitamins as described by Lichter (1982) does not contain Ca(NO3)2.4H2OAdd 347 mg/L Ca(NO3)2.4H2O to the already dissolved mediumSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.43 g/L (w/o calcium nitrate); 1.77g/L (w/ Calcium nitrte)
Catalog No. 50-254-760 Supplier Bioworld Supplier No. 306301032
NLN MediumContains Macronutrients, Micronutrients, and Vitamins as described by Lichter (1982) does not contain Ca(NO3)2.4H2OAdd 347 mg/L Ca(NO3)2.4H2O to the already dissolved mediumSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.43 g/L (w/o calcium nitrate); 1.77g/L (w/ Calcium nitrte)